Cesium hydroxide monohydrate 99.95% (trace metals basis) for analysis

Supplier: Thermo Scientific Chemicals
Danger

18952-0500 AC18952-2500
TS18952-0500EA 219 USD
TS18952-0500 TS18952-2500
Cesium hydroxide monohydrate 99.95% (trace metals basis) for analysis
Cesium hydroxide monohydrate
Formula: HCsO·1H₂O
MW: 167.93 g/mol
Melting Pt: 204.85
MDL Number: MFCD00149664
CAS Number: 35103-79-8
EINECS: 244-344-1
UN: 2682
ADR: 8 II
Merck Index: 12,02055

Order Now

Specification Test Results

Appearance White Crystalline powder
Purity ≥99.95 % (trace metal basis)
Titration with HCl ≥96.5 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤25 ppm
Cesium carbonate (Cs2CO3) ≤5 %
Aluminium (Al) ≤5 ppm
Barium (Ba) ≤15 ppm
Calcium (Ca) ≤2 ppm
Chromium (Cr) ≤1 ppm
Iron (Fe) ≤5 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Potassium (K) ≤150 ppm
Rubidium (Rb) ≤10 ppm
Sodium (Na) ≤75 ppm
Strontium (Sr) ≤5 ppm
Silicon dioxide (SiO2) ≤25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR