Polypropylene glycol, M.W. 2,000

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

19218-0025 19218-5000
TS19218-0025EA 210 USD
TS19218-0025 TS19218-5000
Polypropylene glycol, M.W. 2,000
Polypropylene glycol
Formula: H(OCH₂CH₂CH₂)nOH
Boiling Pt: 200 °C
Melting Pt: –31 °C
Density: 1 g/cm³
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-038-2

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Water ≤0.15 %
Color scale ≤50 APHA
Viscosity 135 to 165 mm²/s (at 40°C)
Acid value ≤0.1 mg KOH/g
Hydroxyl value 53 to 58 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR