Xylitol 99+%

Supplier: Thermo Scientific Chemicals

22598-5000 22598-0025 22598-0250 22598-1000
TS22598-5000EA 90 USD
TS22598-5000 TS22598-0025 TS22598-0250 TS22598-1000
Xylitol 99+%
Xylitol
Formula: C₅H₁₂O₅
Boiling Pt: 215…217 °C
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 96 °C
Water ≤0.5 %
Sulfated ash ≤0.5 %
GC ≥99.0 % (on dry substance)
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤10 ppm
Reducing sugars ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR