ε-Hexalactone monomer 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ε-Caprolactone, Oxepan-2-one, Hexan-6-olide

17344-0010 17344-0025 17344-2500
TS17344-0010EA 151 USD
TS17344-0010 TS17344-0025 TS17344-2500
ε-Hexalactone monomer 99%
ε-Hexalactone
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.03 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003267
CAS Number: 502-44-3
EINECS: 207-938-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR