N-Methyl-N-(trimethylsilyl)trifluoroacetamide 97%

Supplier: Thermo Scientific Chemicals
Warning

22158-0250 22158-1000 AC22158-0050
TS22158-0250EA 245 USD
TS22158-0250 TS22158-1000 TS22158-0050
N-Methyl-N-(trimethylsilyl)trifluoroacetamide 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formula: C₆H₁₂F₃NOSi
Boiling Pt: 130…132 °C (760 mmHg)
Density: 1.07 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR