Tris(trimethylsilyl)silane 96%

Supplier: Thermo Scientific Chemicals
Warning

29106-0250 AC29106-0025
TS29106-0250EA 276 USD
TS29106-0250 TS29106-0025
Tris(trimethylsilyl)silane 96%
Tris(trimethylsilyl)silane
Formula: C₉H₂₈Si₄
Boiling Pt: 82…84 °C (12 mmHg)
Density: 0.8 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00077893
CAS Number: 1873-77-4
UN: 1993
ADR: 3,III
Merck Index: 12,09886

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR