Tetrakis(triphenylphosphine)palladium(0) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
20238-0050
AC20238-0010
AC20238-0250
AC20238-1000
TS20238-0050EA
149
USD
TS20238-0050
TS20238-0010
TS20238-0250
TS20238-1000
Tetrakis(triphenylphosphine)palladium(0) 99%
Tetrakis(triphenylphosphine)palladium(0)
Formula:
C₇₂H₆₀P₄Pd MW: 1155.58 g/mol Melting Pt: 103…107 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 EINECS: 238-086-9 |
Specification Test Results
Appearance | Yellow to yellow-green Powder or crystals |
Infrared spectrum | Conforms |
Carbon (C) | 73.7 to 76.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...