Oil of castor

Supplier: Thermo Scientific Chemicals

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

40416-0025 40416-5000
TS40416-0025EA 125 USD
TS40416-0025 TS40416-5000
Oil of castor
Oil of castor
Boiling Pt: 229 °C (760 mmHg)
Melting Pt: –10 °C
Density: 0.957 g/cm³
Flash Pt: 229 °C
Storage Temperature: Ambient
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
Acid value ≤2 mg KOH/g
Hydroxyl value ≥150 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR