Ethyl methacrylate 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate
21583-0010
21583-5000
TS21583-0010EA
99
USD
TS21583-0010
TS21583-5000
Ethyl methacrylate 98% stabilised
Ethyl methacrylate
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 100 Melting Pt: -48 Density: 0.697 g/cm³ Flash Pt: –9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009161 CAS Number: 97-63-2 EINECS: 209-767-8 UN: 2278 ADR: 3 II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3990 to 1.4010 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | 10 to 120 ppm MEHQ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...