Chromium hexacarbonyl 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl

22105-0100 22105-0500
TS22105-0100EA 150 USD
TS22105-0100 TS22105-0500
Chromium hexacarbonyl 99+%
Chromium hexacarbonyl
Formula: C₆CrO₆
MW: 220.06 g/mol
Boiling Pt: 210
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00010945
CAS Number: 13007-92-6
EINECS: 235-852-4
UN: 3466
ADR: 6.1 III
Merck Index: 13,02253

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR