Maleimide 98%

Supplier: Thermo Scientific Chemicals
Danger

15671-1000 15671-0050 15671-0100 15671-0250 15671-5000
TS15671-1000EA 536 USD
TS15671-1000 TS15671-0050 TS15671-0100 TS15671-0250 TS15671-5000
Maleimide 98%
Maleimide
Formula: C₄H₃NO₂
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR