2,2,2-Trichloroethyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

16181-1000 16181-0250
TS16181-1000EA 122 USD
TS16181-1000 TS16181-0250
2,2,2-Trichloroethyl chloroformate 97%
2,2,2-Trichloroethyl chloroformate
Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 171…172 °C (760 mmHg)
Melting Pt: 1 °C
Density: 1.539 g/cm³
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4720 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR