Cottonseed oil, pure

Supplier: Thermo Scientific Chemicals

17711-0025 17711-5000
TS17711-0025EA 143 USD
TS17711-0025 TS17711-5000
Cottonseed oil, pure
Cottonseed oil
Density: 0.91 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00130872
CAS Number: 8001-29-4
EINECS: 232-280-7

Order Now

Specification Test Results

Appearance Yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4730 (20°C, 589 nm)
Additional info linolenic and higher: ≤1.5%
Acid value ≤0.5 mg KOH/g
Iodine value 95 to 115 g I2/100 g
Saponification value 185 to 198 mg KOH/g
Unsaponifiable matter ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR