tert-Butyl ethyl ether 97%

Supplier: Thermo Scientific Chemicals
Danger

40353-1000 403530250
TS40353-1000EA 526 USD
TS40353-1000 TS403530250
tert-Butyl ethyl ether 97%
Formula: C₆H₁₄O
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.75 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00009225
CAS Number: 637-92-3
EINECS: 211-309-7
UN: 1179
ADR: 3,II
Merck Index: 13,03809

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3746 to 1.3766 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR