Neutral red, pure, certified

Supplier: Thermo Scientific Chemicals

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

41549-0250 41549-1000
TS41549-0250EA 117 USD
TS41549-0250 TS41549-1000
Neutral red, pure, certified
Neutral red
Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥50 %
UV Lambda max 539 to 544 nm
Absorption Ratio P-15/P+15 1 to 1.12
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR