1-Ethyl-3-methylimidazolium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35667-1000
356670050
356670250
TS35667-1000EA
274
USD
TS35667-1000
TS356670050
TS356670250
1-Ethyl-3-methylimidazolium bromide 98%
1-Ethyl-3-methylimidazolium bromide
Formula:
C₆H₁₁BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD03427610 CAS Number: 65039-08-9 |
Specification Test Results
Appearance | Yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...