Benzopinacol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,1,2,2-Tetraphenyl-1,2-ethanediol, 1,1,2,2-Tetraphenylethane-1,2-diol, Tetraphenylethylene glycol, Tetraphenyl-1,2-ethanediol
10559-1000
10559-0100
TS10559-1000EA
170
USD
TS10559-1000
TS10559-0100
Benzopinacol 98%
Benzopinacol
Formula:
(C₆H₅)₂C(OH)C(OH)(C₆H₅)₂ MW: 366.46 g/mol Boiling Pt: 508 °C (1013 hPa) Melting Pt: 184…186 °C |
MDL Number:
MFCD00004448 CAS Number: 464-72-2 EINECS: 207-356-8 Merck Index: 13,01101 |
Specification Test Results
Appearance | White to light yellow Powder or crystals or granules |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...