Methyl (S)-(-)-lactate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

21306-0010 21306-0025 21306-2500
TS21306-0010EA 242 USD
TS21306-0010 TS21306-0025 TS21306-2500
Methyl (S)-(-)-lactate 97%
Methyl (S)-(-)-lactate
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.75 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00064265
CAS Number: 27871-49-4
EINECS: 203-968-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4284 to 1.4304 (20°C, 589 nm)
GC ≥99.8 %
Solubility (In water) Passes test
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds Each of acetone, formaldehyde and acetaldehyde
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR