Quinhydrone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)

41877-1000 41877-0250 41877-5000
TS41877-1000EA 106 USD
TS41877-1000 TS41877-0250 TS41877-5000
Quinhydrone 98%
Quinhydrone
Formula: C₆H₄(OH)₂·C₆H₄O₂
MW: 218.21 g/mol
Melting Pt: 168…172 °C
Density: 1.4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010310
CAS Number: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,08148

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C (Dec)
Titration Iodometric ≥97.5 %
Sulfated ash ≤0.2 %
HPLC 48.0 to 52.0 % Hydroquinone
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR