1,4-Butanesultone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Butane sultone

10767-1000 10767-5000 10767-0250
TS10767-1000EA 133 USD
TS10767-1000 TS10767-5000 TS10767-0250
1,4-Butanesultone 99%
1,4-Butanesultone
Formula: C₄H₈O₃S
MW: 136.17
Boiling Pt: 153
Melting Pt: –23 °C
Density: 0.81 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00006584
CAS Number: 1633-83-6
EINECS: 210-971-4
UN: 2282
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4125 to 1.4155 (20°C, 589 nm)
GC ≥99.0 %
Titration with TBAH ≤0.1 % 4-Hydoxybutanesulfonic acid
Chloride (Cl) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR