Ammonium molybdate(VI) 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate

44854-1000 AC44854-0050 AC44854-0250
TS44854-1000EA 736 USD
TS44854-1000 TS44854-0050 TS44854-0250
Ammonium molybdate(VI) 99.99% (trace metals basis)
Ammonium molybdate(VI)
Formula: H₈MoN₂O₄
MW: 196.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011596
CAS Number: 13106-76-8

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Total trace metal impurities ≤100 ppm
Molybdenum (Mo) 47.0 to 50.7 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR