Benzethonium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10538-5000
10538-0025
10538-1000
TS10538-5000EA
212
USD
TS10538-5000
TS10538-0025
TS10538-1000
Benzethonium chloride 97%
Benzethonium chloride
Formula:
C₂₇H₄₂ClNO₂ Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011742 CAS Number: 121-54-0 EINECS: 204-479-9 UN: 2923 ADR: 8,III Merck Index: 13,01074 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 158 to 163 °C |
Titration Argentometric | ≥96.0 % (on dry substance) |
Loss on drying | ≤5 % (105 °C, 4 hrs) |
Zinc (Zn) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...