Tetrabutylammonium hexafluorophosphate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35258-1000
AC35258-0250
AC35258-5000
TS35258-1000EA
216
USD
TS35258-1000
TS35258-0250
TS35258-5000
Tetrabutylammonium hexafluorophosphate 98%
Tetrabutylammonium hexafluorophosphate
Formula:
C₁₆H₃₆F₆NP MW: 387.43 g/mol Melting Pt: 244…246 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011748 CAS Number: 3109-63-5 EINECS: 221-472-6 |
Specification Test Results
Appearance | White to off-white Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 242 to 247 °C |
Elemental Analysis | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...