1-Pentanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
39665-1000
AC39665-0250
AC39665-5000
TS39665-1000EA
954
USD
TS39665-1000
TS39665-0250
TS39665-5000
1-Pentanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
1-Pentanesulphonic acid sodium salt
Formula:
C₅H₁₁O₃SNa MW: 174.2 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (150°C) |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...