Silver diethyldithiocarbamate 99% for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver
11429-1000
AC11429-0050
AC11429-0250
TS11429-1000EA
814
USD
TS11429-1000
TS11429-0050
TS11429-0250
Silver diethyldithiocarbamate 99% for analysis
Silver diethyldithiocarbamate
Formula:
(C₂H₅)₂NCSSAg MW: 256.14 g/mol Melting Pt: 172…175 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004929 CAS Number: 1470-61-7 EINECS: 216-003-7 |
Specification Test Results
Appearance | Green to khaki Powder |
Infrared spectrum | Conforms |
Solubility | (0.5% in pyridine) clear |
Titration with NH4SCN | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...