2-Methoxy-1-methylethyl acetate (PGMEA) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA

41390-0010 41390-0050
TS41390-0010EA 84 USD
TS41390-0010 TS41390-0050
2-Methoxy-1-methylethyl acetate (PGMEA) 99%
2-Methoxy-1-methylethyl acetate (PGMEA)
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145.8 °C (1013 hPa)
Melting Pt: –87 °C
Density: 0.96 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00038500
CAS Number: 108-65-6
EINECS: 203-603-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
Water ≤0.05 %
GC ≥98.5 %
Acidity (CH3COOH) ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR