Triacetin (glycerol triacetate) 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
13922-5000
13922-0010
13922-0025
13922-0100
TS13922-5000EA
74
USD
TS13922-5000
TS13922-0010
TS13922-0025
TS13922-0100
Triacetin (glycerol triacetate) 99%
Triacetin (glycerol triacetate)
Formula:
C₉H₁₄O₆ MW: 218.21 g/mol Boiling Pt: 258 °C (760 mmHg) Melting Pt: 3 °C Density: 1.155 g/cm³ Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008716 CAS Number: 102-76-1 EINECS: 203-051-9 Merck Index: 13,09664 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4330 (20°C, 589 nm) |
Density | (25°C) 1.154 to 1.158 g/ml |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...