Tributyrin (glycerol tributyrate) 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate

15088-0010 15088-1000 15088-2500
TS15088-0010EA 165 USD
TS15088-0010 TS15088-1000 TS15088-2500
Tributyrin (glycerol tributyrate) 97%
Tributyrin (glycerol tributyrate)
Formula: C₁₅H₂₆O₆
MW: 302.37 g/mol
Boiling Pt: 305…310 °C (760 mmHg)
Melting Pt: –75 °C
Density: 1.0335 g/cm³
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00009392
CAS Number: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4380 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR