Gallium trinitrate hydrate 99.9998% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

21244-0250 AC21244-0010 AC21244-0050 AC21244-1000
TS21244-0250EA 231 USD
TS21244-0250 TS21244-0010 TS21244-0050 TS21244-1000
Gallium trinitrate hydrate 99.9998% (trace metals basis)
Gallium trinitrate hydrate
Formula: GaN₃O₉·H₂O
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.0002 %
Gallium (Ga) 18 to 22 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR