Tetrabutylammonium fluoride 1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

43351-1000 43351-8000
TS43351-1000EA 86 USD
TS43351-1000 TS43351-8000
Tetrabutylammonium fluoride 1 M in THF, AcroSeal™
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Density: 0.887 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011747
CAS Number: 429-41-4
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to amber to light green to brown Solution
Water ≤8 %
Titration with HClO4 0.90 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR