Diisobutylaluminium hydride 1 M in hexane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

18379-1000 18379-4000 183798000
TS18379-1000EA 86 USD
TS18379-1000 TS18379-4000 TS183798000
Diisobutylaluminium hydride 1 M in hexane, AcroSeal™
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: –70 °C
Density: 0.701 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2,I

Order Now

Specification Test Results

Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)
Melting point 118 to 123 °C
Loss on drying ≤0.5 %
Titration with HClO4 98 to 102 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR