Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di-i-butylaluminum hydride, DIBAL-H
20105-1000
20105-8000
TS20105-1000EA
80
USD
TS20105-1000
TS20105-8000
Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal™
Diisobutylaluminium hydride
Formula:
C₈H₁₉Al MW: 142.22 g/mol Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008928 CAS Number: 1191-15-7 EINECS: 214-729-9 UN: 3399 ADR: 4.3,I |
Specification Test Results
Appearance | Clear solution |
Strength | 19.5 to 20.5 % (DIBAL-H, on Al basis) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...