Diisobutylaluminium hydride 1.1 M in cyclohexane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

20103-1000 201038000
TS20103-1000EA 78 USD
TS20103-1000 TS201038000
Diisobutylaluminium hydride 1.1 M in cyclohexane, AcroSeal™
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22
Melting Pt: -80
Density: 0.798
Flash Pt: –018 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR