Potassium tetrachloroplatinate(II) Pt 46-47%

Supplier: Thermo Scientific Chemicals
Danger

19536-0050 AC19536-0010 AC19536-0250
TS19536-0050EA 547 USD
TS19536-0050 TS19536-0010 TS19536-0250
Potassium tetrachloroplatinate(II) Pt 46-47%
Potassium tetrachloroplatinate(II)
Formula: Cl₄K₂Pt
Boiling Pt: 35…40 °C (20 mmHg)
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,07855

Order Now

Specification Test Results

Appearance Red-brown Crystalline powder or crystals
Total trace metal impurities ≤500 ppm (referring to Pt)
Gravimetry 46 to 47 % (Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR