Phenylacetylene 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

15246-5000 15246-0250 15246-1000
TS15246-5000EA 542 USD
TS15246-5000 TS15246-0250 TS15246-1000
Phenylacetylene 98%, pure
Phenylacetylene
Formula: C₈H₆
MW: 102.14 g/mol
Boiling Pt: 266…283 °C (760 mmHg)
Melting Pt: –7 °C
Density: 0.84 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 203-984-1
UN: 1760
ADR: 8,III
Merck Index: 13,03892

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤20 APHA
Residue after reduction ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR