Styrene 99.5% stabilized for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylethene, Phenylethylene, Vinylbenzene

22053-0010 22053-2500
TS22053-0010EA 57 USD
TS22053-0010 TS22053-2500
Styrene 99.5% stabilized for analysis
Styrene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 145.2 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.906 g/cm³
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008612
CAS Number: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3,III
Merck Index: 13,08944

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5480 (20 °C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA
Stabilizer 10 to 20 ppm p-tert-Butylcatechol
Aldehyde ≤0.02 % (as benzaldehyde)
Polymer ≤10 ppm
Peroxides ≤0.01 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR