Lithium dodecyl sulphate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
41330-1000
AC41330-0250
TS41330-1000EA
433
USD
TS41330-1000
TS41330-0250
Lithium dodecyl sulphate 99+%
Lithium dodecyl sulphate
Formula:
C₁₂H₂₅LiO₄S MW: 272.34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007467 CAS Number: 2044-56-6 EINECS: 218-058-2 UN: 1325 ADR: 4.1 III |
Specification Test Results
Appearance | White Glistening crystalline powder or flakes |
Infrared spectrum | Conforms |
UV | at 280 nm A: ≤0.1 |
Titration after Ion exchange | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...