Trimethylsilyl trifluoromethanesulphonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylsilyl trifluoromethanesulphonate
20944-0500
AC20944-0100
AC20944-2500
TS20944-0500EA
210
USD
TS20944-0500
TS20944-0100
TS20944-2500
Trimethylsilyl trifluoromethanesulphonate 99%
Trimethylsilyl trifluoromethanesulphonate
Formula:
C₄H₉F₃O₃SSi MW: 222.26 g/mol Boiling Pt: 77 Density: 1.225 g/cm³ Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000406 CAS Number: 27607-77-8 EINECS: 248-565-4 UN: 2920 ADR: 8 II Merck Index: 12,09849 |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3600 to 1.3620 (20°C, 589 nm) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...