4'-Chloroacetanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloroacetanilide, Paracetamol impurity J, N-(4-Chlorophenyl)acetamide

16995-0500 16995-0050 16995-2500
TS16995-0500EA 84 USD
TS16995-0500 TS16995-0050 TS16995-2500
4'-Chloroacetanilide 97%
4'-Chloroacetanilide
Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 237 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.99 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 214-735-1
UN: 3334
ADR: 9,III
Merck Index: 13,02128

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5150 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +16° to +18° (20°C, 589 nm) (c=1,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR