Mercury(II) nitrate monohydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate

42394-5000 423941000
TS42394-5000EA 513 USD
TS42394-5000 TS423941000
Mercury(II) nitrate monohydrate ACS
Mercury(II) nitrate monohydrate
Formula: Hg(NO₃)₂·1H₂O
MW: 342.62 g/mol
Melting Pt: 79
Density: 4.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149736
CAS Number: 7783-34-8
EINECS: 233-152-3
UN: 1625
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Titration with NH4SCN ≥98.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤10 ppm
Residue after reduction ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR