Mercury(I) nitrate dihydrate 98% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimercury dinitrate dihydrate, Mercurous nitrate dihydrate

21313-0500 AC21313-0010 AC21313-2500
TS21313-0500EA 103 USD
TS21313-0500 TS21313-0010 TS21313-2500
Mercury(I) nitrate dihydrate 98% for analysis
Mercury(I) nitrate dihydrate
Formula: Hg₂N₂O₆·2H₂O
MW: 561.22 g/mol
Melting Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00150812
CAS Number: 14836-60-3
EINECS: 233-886-4
UN: 1627
ADR: 6.1 II
Merck Index: 13,05923

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Assay ≥97.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Mercuric Mercury (as Hg) ≤0.5 %
Insoluble matter ≤0.005 % (in dilute HNO3)
Residue after reduction ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR