tetra-Chloroauric(III) acid trihydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

41107-0010 41107-0250 AC41107-0050
TS41107-0010EA 186 USD
TS41107-0010 TS41107-0250 TS41107-0050
tetra-Chloroauric(III) acid trihydrate ACS
tetra-Chloroauric(III) acid trihydrate
Formula: HAuCl₄·3H₂O
MW: 393.83 g/mol
Melting Pt: 30
Storage Temperature: Refrigerator
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance Yellow to orange Powder and/or chunks (May have a brown hue)
Assay ≥49.0 % (Au)
Insoluble matter ≤0.1 % (10 % in ether)
Alkalies+oth.metals (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR