trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Titriplex® IV

15500-1000 15500-5000 15500-0025 15500-0250
TS15500-1000EA 215 USD
TS15500-1000 TS15500-5000 TS15500-0025 TS15500-0250
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula: C₁₄H₂₂N₂O₈·1H₂O
Melting Pt: >210 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 97.5 to 102.5 %
Loss on drying 4.0 to 6.9 % (1g,150°C,6h)
Sulfated ash ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR