N-(1-Naphthyl)ethylenediamine dihydrochloride 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

42399-1000 42399-5000 42399-0050 42399-0250
TS42399-1000EA 466 USD
TS42399-1000 TS42399-5000 TS42399-0050 TS42399-0250
N-(1-Naphthyl)ethylenediamine dihydrochloride 98+% ACS
N-(1-Naphthyl)ethylenediamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to beige to pink Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 % (on dry substance)
Water ≤5 % (K.F.)
Loss on drying ≤5 % (1 g, 80°C, 4 h)
Solubility Passes test
Sensitivity (to sulfanilamide) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR