Starch, soluble ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Starch, Vitex® iodine indicator, Amylodextrin
42449-5000
42449-0020
42449-0100
42449-1000
TS42449-5000EA
92
USD
TS42449-5000
TS42449-0020
TS42449-0100
TS42449-1000
Starch, soluble ACS
Starch, soluble
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Melting Pt: 256…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-84-9 EINECS: 232-686-4 Merck Index: 13,08878 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Solubility | Passes test |
Residue after ignition | ≤0.4 % |
pH | 5.0 to 7.0 (2 % solution at 25.0°C) |
Sensitivity | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...