Benzalkonium chloride alkyl distribution from C8H17 to C16H33

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

21541-0010 21541-0051 21541-1000
TS21541-0010EA 272 USD
TS21541-0010 TS21541-0051 TS21541-1000
Benzalkonium chloride alkyl distribution from C8H17 to C16H33
Benzalkonium chloride
Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Melting Pt: 40 °C
Density: 0.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to white to yellow Solid or thick gel (May contain yellowish-white fragments)
Infrared spectrum Conforms
Assay 95.0 to 104.0 % (on anhydrous basis)
Water ≤10.0 % (K.F.)
Sulfated ash ≤0.5 %
Total amines ≤3.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR