Zinc sulphate monohydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

38980-2500 AC38980-0010 AC38980-0050
TS38980-2500EA 45 USD
TS38980-2500 TS38980-0010 TS38980-0050
Zinc sulphate monohydrate 99%
Zinc sulphate monohydrate
Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
UN: 3077
ADR: 9 III
Merck Index: 13,10213

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR