Vanadium(III) chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

19700-0250 AC19700-0050 AC19700-1000
TS19700-0250EA 158 USD
TS19700-0250 TS19700-0050 TS19700-1000
Vanadium(III) chloride 97%
Vanadium(III) chloride
Formula: Cl₃V
MW: 157.3 g/mol
Melting Pt: 250 °C (dec.)
Density: 3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011454
CAS Number: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8,III

Order Now

Specification Test Results

Appearance Dark blue to violet to black Powder
Titration Argentometric ≥96.0 %
Insoluble matter ≤0.5 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR