Titanium(IV) oxide, Aeroxide® P25

Supplier: Thermo Scientific Chemicals

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

38429-0010 AC38429-0500 AC38429-2500
TS38429-0010EA 303 USD
TS38429-0010 TS38429-0500 TS38429-2500
Titanium(IV) oxide, Aeroxide® P25
Titanium(IV) oxide
Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900
Melting Pt: 1855
Density: 4.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Specification Test Results

Appearance White Powder
Assay ≥99.5 % (on ignited substance) Typical
Specific surface area 35 to 65 m²/g
pH 3.5 to 4.5 (4 % dispersion)
Acidity (HCl) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR