Tetramethylammonium hydroxide pentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

42053-1000 AC42053-0050 AC42053-0250 AC42053-5000
TS42053-1000EA 273 USD
TS42053-1000 TS42053-0050 TS42053-0250 TS42053-5000
Tetramethylammonium hydroxide pentahydrate 98%
Tetramethylammonium hydroxide pentahydrate
Formula: C₄H₁₃NO·5H₂O
Melting Pt: 62…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 73 °C
Titration with HCl 97.5 to 103 %
Carbonate (CO3) ≤7500 ppm
Iron (Fe) ≤5 ppm
Sodium + Potassium (Na+K) ≤5 ppm
Impurity ≤0.1 % Halides (as Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR